Skip to Content
Merck
All Photos(1)

Documents

V800234

Sigma-Aldrich

Lead(IV) oxide

LR, ≥94%

Synonym(s):

Lead (su)peroxide, Lead dioxide, Lead peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbO2
CAS Number:
Molecular Weight:
239.20
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

LR

product line

Vetec

Assay

≥94%

form

powder

SMILES string

O=[Pb]=O

InChI

1S/2O.Pb

InChI key

YADSGOSSYOOKMP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Ox. Sol. 3 - Repr. 1A - STOT RE 2

Target Organs

Blood,Central nervous system,Immune system,Kidney

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service