Skip to Content
Merck

1707792

USP

Ursodiol Related Compound H

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3β, 7β-Dihydroxy-5beta-cholan-24-oic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

25 MG

CZK 31,600.00

CZK 31,600.00


Please contact Customer Service for Availability

Request a Bulk Order

About This Item

Empirical Formula (Hill Notation):
C24H40O4
CAS Number:
Molecular Weight:
392.57
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

SMILES string

O[C@@H]1[C@@H]2[C@@H]([C@@]4([C@H](C1)C[C@H](CC4)O)C)CC[C@]3([C@H]2CC[C@@H]3[C@@H](CCC(=O)O)C)C

InChI

1S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16+,17-,18+,19+,20+,22+,23+,24-/m1/s1

InChI key

RUDATBOHQWOJDD-DNMBCGTGSA-N

API family

ursodiol

packaging

pkg of 25 mg

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
161588712640131604585
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

packaging

pkg of 25 mg

packaging

-

packaging

pkg of 50 mg

packaging

pkg of 20 mg

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

API family

ursodiol

API family

sorafenib

API family

ethosuximide

API family

rivaroxaban

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Ursodiol Related Compound H USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  

Other Notes

Sales restrictions may apply.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service