Skip to Content
Merck

Skip To

1652500

USP

Thalidomide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(±)-Thalidomide, (±)-2-(2,6-Dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
13 000,00 CZK

About This Item

Empirical Formula (Hill Notation):
C13H10N2O4
CAS Number:
Molecular Weight:
258.23
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

13 000,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

thalidomide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C1CCC(N2C(=O)c3ccccc3C2=O)C(=O)N1

InChI

1S/C13H10N2O4/c16-10-6-5-9(11(17)14-10)15-12(18)7-3-1-2-4-8(7)13(15)19/h1-4,9H,5-6H2,(H,14,16,17)

InChI key

UEJJHQNACJXSKW-UHFFFAOYSA-N

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thalidomide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Thalidomide Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR324910295451231502
description

United States Pharmacopeia (USP) Reference Standard

description

pharmaceutical secondary standard, certified reference material

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

neat

format

neat

API family

thalidomide

API family

thalidomide

API family

amlodipine

API family

dyphylline

Gene Information

human ... CRBN(51185), CUL4A(8451), DDB1(1642), RBX1(9978)

Gene Information

-

Gene Information

-

Gene Information

human ... PDE3A(5139), PDE4A(5141), PDE4B(5142), PDE4C(5143), PDE4D(5144)


Still not finding the right product?

Explore all of our products under Thalidomide


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters