Skip to Content
Merck

Skip To

1638000

USP

Sulfisoxazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4-Amino-N-(3,4-dimethyl-5-isoxazolyl)benzenesulfonamide, N1-(3,4-Dimethyl-5-isoxazolyl)sulfanilamide, Sulfafurazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
10 600,00 CZK

About This Item

Empirical Formula (Hill Notation):
C11H13N3O3S
CAS Number:
Molecular Weight:
267.30
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
6737262
eCl@ss:
25140904

10 600,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

sulfisoxazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)
pharmaceutical

format

neat

SMILES string

Cc1noc(NS(=O)(=O)c2ccc(N)cc2)c1C

InChI

1S/C11H13N3O3S/c1-7-8(2)13-17-11(7)14-18(15,16)10-5-3-9(12)4-6-10/h3-6,14H,12H2,1-2H3

InChI key

NHUHCSRWZMLRLA-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S1975000317391632004
description

United States Pharmacopeia (USP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

description

VETRANAL®, analytical standard

description

United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

analytical standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)
pharmaceutical

application(s)

pharmaceutical
pharmaceutical (small molecule)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

agency

USP

agency

-

agency

-

agency

USP

API family

sulfisoxazole

API family

sulfafurazole

API family

-

API family

sulfanilamide


Still not finding the right product?

Explore all of our products under Sulfisoxazole


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters