Skip to Content
Merck

Skip To

1500003

USP

Paromomycin sulfate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Paromomycin sulfate salt, Aminosidine sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
125 mg
Please contact Customer Service for Availability
CZK 9,460.00

About This Item

Empirical Formula (Hill Notation):
C23H45N5O14 · xH2SO4
CAS Number:
Molecular Weight:
615.63 (free base basis)
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 9,460.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

paromomycin

form

solid

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

[S](=O)(=O)(O)O.N[C@H]1[C@H](O[C@H]([C@H]([C@@H]1O)O)CN)O[C@@H]2[C@H](OC([C@@H]2O)O[C@@H]3[C@H]([C@@H](C[C@@H](C3O[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4N)O)O)CO)N)N)O)CO

InChI

1S/C23H45N5O14.H2O4S/c24-2-7-13(32)15(34)10(27)21(37-7)41-19-9(4-30)39-23(17(19)36)42-20-12(31)5(25)1-6(26)18(20)40-22-11(28)16(35)14(33)8(3-29)38-22;1-5(2,3)4/h5-23,29-36H,1-4,24-28H2;(H2,1,2,3,4)/t5-,6+,7+,8-,9-,10-,11-,12+,13-,14-,15-,16-,17-,18?,19-,20-,21-,22-,23?;/m1./s1

InChI key

LJRDOKAZOAKLDU-UMIPPVEZSA-N

General description

Paromomycin sulfate is an aminoglycoside antibiotic primarily used in the treatment of parasitic infections and certain bacterial infections. It inhibits protein synthesis at the 30S ribosomal subunit. The USP reference standard is employed in potency and identity assays.

The USP biologics antibiotics category includes a wide range of antimicrobial agents that are essential in treating bacterial infections. These antibiotics are derived from various natural sources or synthesized to combat specific pathogens effectively. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of antibiotic therapeutics throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Paromomycin Sulfate USP reference standard is used to prepare standard solutions for the identification and assay of paromomycin sulfate.
Also, for use with USP monographs such as:
  • Paromomycin Sulfate Capsules
  • Paromomycin Oral Solution

Other Notes

Sales restrictions may apply.
This product is part of the USP Biologics program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
109100616128011148001
manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

form

solid

form

solid

form

solid

form

solid

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

paromomycin

API family

capreomycin

API family

sisomicin

API family

colistin


Still not finding the right product?

Explore all of our products under Paromomycin sulfate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters