Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
200 mg | Please contact Customer Service for Availability | CZK 18,000.00 |
About This Item
Empirical Formula (Hill Notation):
C15H14N2O2
CAS Number:
Molecular Weight:
254.28
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
grade
pharmaceutical primary standard
agency
USP
API family
nepafenac
packaging
pkg of 200 mg
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
NC(=O)Cc1cccc(c1N)C(=O)c2ccccc2
InChI
1S/C15H14N2O2/c16-13(18)9-11-7-4-8-12(14(11)17)15(19)10-5-2-1-3-6-10/h1-8H,9,17H2,(H2,16,18)
InChI key
QEFAQIPZVLVERP-UHFFFAOYSA-N
General description
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Application
Nepafenac USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Biochem/physiol Actions
Nepafenac is a NSAID (nonsteroidal anti inflammatory drug) that is routinely used in opthamology to control pain following cataract surgery.
Nepafenac is a NSAID (nonsteroidal anti inflammatory drug) used in opthamology.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
1 of 1
This Item | |||
|---|---|---|---|
| application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecules) | application(s) - | application(s) pharmaceutical (small molecule) |
| format neat | format neat | format - | format neat |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. room temp | storage temp. 2-8°C |
| manufacturer/tradename USP | manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP |
| packaging pkg of 200 mg | packaging - | packaging - | packaging pkg of 20 mg |
| API family nepafenac | API family nepafenac | API family - | API family nicardipine |
Still not finding the right product?
Explore all of our products under Nepafenac
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


