Skip to Content
Merck

Skip To

1457902

USP

Nefazodone hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-5-ethyl- 2,4-dihydro-4-(2-phenoxyethyl)-3H-1,2,4-triazol-3-one hydrochloride, BMY-13754, MJ-13754-1, Serzone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
26 600,00 CZK

About This Item

Empirical Formula (Hill Notation):
C25H32ClN5O2 · HCl
CAS Number:
Molecular Weight:
506.47
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

26 600,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

nefazodone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O=C1N(CCCN2CCN(C3=CC=CC(Cl)=C3)CC2)N=C(CC)N1CCOC4=CC=CC=C4.Cl

InChI

1S/C25H32ClN5O2.ClH/c1-2-24-27-31(25(32)30(24)18-19-33-23-10-4-3-5-11-23)13-7-12-28-14-16-29(17-15-28)22-9-6-8-21(26)20-22;/h3-6,8-11,20H,2,7,12-19H2,1H3;1H

InChI key

DYCKFEBIOUQECE-UHFFFAOYSA-N

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.

Application

Nefazodone hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Nefazodone Hydrochloride Tablets

Biochem/physiol Actions

Novel antidepressant; mixed 5-HT2A serotonin receptor antagonist/serotonin uptake inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
N553614579241673533
description

United States Pharmacopeia (USP) Reference Standard

description

≥98% (HPLC), solid

description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

format

neat

format

-

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

Gene Information

human ... HTR2A(3356), HTR2C(3358), SLC6A2(6530), SLC6A4(6532)

Gene Information

human ... HTR2A(3356), HTR2C(3358), SLC6A2(6530), SLC6A4(6532)

Gene Information

-

Gene Information

-

API family

nefazodone

API family

-

API family

nefazodone

API family

trazodone


Still not finding the right product?

Explore all of our products under Nefazodone hydrochloride


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters