Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
methylphenidate
manufacturer/tradename
USP
drug control
USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal), kontrollierte Droge in Deutschland
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monographs such as:
- Methylphenidate Hydrochloride
- Methylphenidate Hydrochloride Extended-Release Tablets
Analysis Note
Other Notes
Legal Information
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.
English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Storage Class
3 - Flammable liquids
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
target_organs
Eyes,Central nervous system
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
67-56-1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


