Skip to Content
Merck

Skip To

1377009

USP

Meclizine hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Meclizine dihydrochloride monohydrate, 1-([4-Chlorophenyl]phenylmethyl)-4-([3-methylphenyl]methyl)piperazine dihydrochloride monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Please contact Customer Service for Availability
11 500,00 CZK

About This Item

Empirical Formula (Hill Notation):
C25H27ClN2 · 2HCl · H2O
CAS Number:
Molecular Weight:
481.89
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

11 500,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

meclizine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].[Cl-].Clc1ccc(cc1)C(N3CCN(CC3)Cc4cc(ccc4)C)c2ccccc2.O.[H+].[H+]

InChI

1S/C25H27ClN2.2ClH.H2O/c1-20-6-5-7-21(18-20)19-27-14-16-28(17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23;;;/h2-13,18,25H,14-17,19H2,1H3;2*1H;1H2

InChI key

KDLHYOMCWBWLMM-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meclizine hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1377029M0220000BP220
description

United States Pharmacopeia (USP) Reference Standard

description

United States Pharmacopeia (USP) Reference Standard

description

European Pharmacopoeia (EP) Reference Standard

description

British Pharmacopoeia (BP) Reference Standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

format

neat

format

neat

format

neat

API family

meclizine

API family

meclizine

API family

meclozine

API family

meclozine

agency

USP

agency

USP

agency

-

agency

BP


Still not finding the right product?

Explore all of our products under Meclizine hydrochloride


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters