Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C3H2ClF5O
CAS Number:
Molecular Weight:
184.49
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Product 1349003 is not currently sold in your country. Contact Technical Service
grade
pharmaceutical primary standard
agency
USP
API family
isoflurane
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
FC(F)(F)C(Cl)OC(F)F
InChI
1S/C3H2ClF5O/c4-1(3(7,8)9)10-2(5)6/h1-2H
InChI key
PIWKPBJCKXDKJR-UHFFFAOYSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Isoflurane USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Biochem/physiol Actions
Isoflurane is a tandem pore potassium channel activator.
Isoflurane is a tandem pore potassium channel activator. It is also a very widely used anesthetic for animal research and for in vitro studies on anesthesia mechanisms.
Analysis Note
These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.
Other Notes
Sales restrictions may apply.
Still not finding the right product?
Explore all of our products under Isoflurane
signalword
Warning
hcodes
Hazard Classifications
STOT SE 3
target_organs
Central nervous system
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
