Skip to Content
Merck

Skip To

1311408

USP

Homosalate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

3,3,5-Trimethylcyclohexyl salicylate, 2-Hydroxybenzoic acid 3,3,5-trimethylcyclohexyl ester, Homomenthyl salicylate, Homosalate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Please contact Customer Service for Availability
CZK 10,600.00

About This Item

Empirical Formula (Hill Notation):
C16H22O3
CAS Number:
Molecular Weight:
262.34
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2731604

CZK 10,600.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

homosalate

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1CC(CC(C)(C)C1)OC(=O)c2ccccc2O

InChI

1S/C16H22O3/c1-11-8-12(10-16(2,3)9-11)19-15(18)13-6-4-5-7-14(13)17/h4-7,11-12,17H,8-10H2,1-3H3

InChI key

WSSJONWNBBTCMG-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Homosalate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1477943PHR10851477900
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

API family

homosalate

API family

octisalate

API family

homosalate

API family

octinoxate

storage temp.

2-8°C

storage temp.

-

storage temp.

2-30°C

storage temp.

-


Still not finding the right product?

Explore all of our products under Homosalate


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

339.4 °F - closed cup

flash_point_c

170.8 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters