Skip to Content
Merck

Skip To

1288816

USP

Genistein

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4′,5,7-Trihydroxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
15 mg
Please contact Customer Service for Availability
CZK 9,050.00

About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
263823

CZK 9,050.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

genistein

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

Oc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18H

InChI key

TZBJGXHYKVUXJN-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Genistein USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Powdered Red Clover
  • Powdered Red Clover Extract
  • Powdered Soy Isoflavones Extract
  • Red Clover
  • Red Clover Aerial Parts Isoflavone Aglycones Dry Extract
  • Red Clover Tablets
  • Soy Isoflavones Capsules

Biochem/physiol Actions

Inhibitor of tyrosine protein kinase; competitive inhibitor of ATP in other protein kinase reactions. Antiangiogenic agent, down-regulates the transcription of genes involved in controlling angiogenesis.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR18590536059092136
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

primary reference standard

grade

analytical standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

HWI

manufacturer/tradename

-

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

food and beverages

application(s)

food and beverages

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

2-8°C

API family

genistein

API family

genistein

API family

-

API family

-


Still not finding the right product?

Explore all of our products under Genistein


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters