Skip to Content
Merck

Skip To

1276001

USP

Fluocinonide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Fluocinonide, (6α,11β,16α)-21-(acetyloxy)-6,9-difluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-pregna-1,4-diene-3,20-dione, Lonide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
200 mg
Please contact Customer Service for Availability
CZK 10,900.00

About This Item

Empirical Formula (Hill Notation):
C26H32F2O7
CAS Number:
Molecular Weight:
494.52
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 10,900.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

fluocinonide

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(=O)OCC(=O)[C@@]12OC(C)(C)O[C@@H]1C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]23C

InChI

1S/C26H32F2O7/c1-13(29)33-12-20(32)26-21(34-22(2,3)35-26)10-15-16-9-18(27)17-8-14(30)6-7-23(17,4)25(16,28)19(31)11-24(15,26)5/h6-8,15-16,18-19,21,31H,9-12H2,1-5H3/t15-,16-,18-,19-,21+,23-,24-,25-,26+/m0/s1

InChI key

WJOHZNCJWYWUJD-IUGZLZTKSA-N

Gene Information

human ... NR3C1(2908)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fluocinonide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Fluocinonide Ointment
  • Fluocinonide Cream
  • Fluocinonide Gel
  • Fluocinonide Topical Solution

Biochem/physiol Actions

Flucinonide is a fluorinated glucocorticid that is used as a topical anti-inflammatory agent. Recently, flucinonide and other fluorinated glucocorticolids were identified as smoothend agonists. Flucinonide induces expression of Gli-reporter luciferase in Shh-LIGHT2 cells.
very potent corticosteroid

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP489SML00991275009
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

-

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

-

format

neat

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

API family

fluocinonide

API family

fluocinonide

API family

-

API family

fluocinolone


Still not finding the right product?

Explore all of our products under Fluocinonide


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral - Repr. 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters