Skip to Content
Merck

Skip To

1192808

USP

Diethanolamine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
605315
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

API family

diethanolamine

autoignition temp.

689 °F

expl. lim.

10.6 %

manufacturer/tradename

USP

refractive index

n20/D 1.477 (lit.)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diethanolamine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
3981791.162058.03116
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

ACS reagent

grade

reagent grade

grade

synthesis grade

format

neat

format

-

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

-

mp

28 °C (lit.)

mp

28 °C (lit.)

mp

28 °C

mp

28 °C

vapor density

3.6 (vs air)

vapor density

3.6 (vs air)

vapor density

-

vapor density

-


Still not finding the right product?

Explore all of our products under Diethanolamine


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

target_organs

Kidney,Liver,Blood

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters