Skip to Content
Merck

Skip To

1171207

USP

23-EPI-26-Deoxyactein

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(3β,12β,16β,23S,24R,25R)-12-(Acetyloxy)-16,23:23,26:24,25-triepoxy-9,19-cyclolanostan-3-yl-β-D-xylopyranoside, 27-Deoxyacetein, 27-Deoxyacetylacetol-O-β-D-xylopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 mg
Please contact Customer Service for Availability
43 300,00 CZK

About This Item

Empirical Formula (Hill Notation):
C37H56O10
CAS Number:
Molecular Weight:
660.83
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904

43 300,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

actein

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H](C1)O)O)O)O[C@@H]2C([C@H]3[C@@]4([C@]5([C@H]([C@]6([C@@]([C@@H]7[C@@H](O[C@@]8(OC[C@]9(O[C@H]98)C)C[C@H]7C)C6)([C@@H](C5)OC(=O)C)C)C)CC3)C4)CC2)(C)C

InChI

1S/C37H56O10/c1-18-12-37(30-33(6,47-30)17-43-37)46-21-13-32(5)23-9-8-22-31(3,4)24(45-29-28(41)27(40)20(39)15-42-29)10-11-35(22)16-36(23,35)14-25(44-19(2)38)34(32,7)26(18)21/h18,20-30,39-41H,8-17H2,1-7H3/t18-,20-,21+,22+,23+,24+,25-,26+,27+,28-,29+,30-,32+,33-,34-,35-,36+,37+/m1/s1

InChI key

GCMGJWLOGKSUGX-RBKCHLQLSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

23-EPI-26-Deoxyactein USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Black Cohosh
  • Powdered Black Cohosh Extract
  • Black Cohosh Fluidextract
  • Powdered Black Cohosh
  • Black Cohosh Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters