Skip to Content
Merck

Skip To

1119309

USP

Chlorthalidone Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

4′-Chloro-3′-sulfamoyl-2-benzophenone-2-carboxylic acid, 2-[3-(Aminosulfonyl)-4-chlorobenzoyl]benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
28 800,00 CZK

About This Item

Empirical Formula (Hill Notation):
C14H10ClNO5S
CAS Number:
Molecular Weight:
339.75
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
eCl@ss:
25140904

28 800,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

chlorthalidone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)
pharmaceutical

format

neat

storage temp.

−20°C

SMILES string

[S](=O)(=O)(N)c1c(ccc(c1)C(=O)c2c(cccc2)C(=O)O)Cl

InChI

1S/C14H10ClNO5S/c15-11-6-5-8(7-12(11)22(16,20)21)13(17)9-3-1-2-4-10(9)14(18)19/h1-7H,(H,18,19)(H2,16,20,21)

InChI key

UKRADWWBRBBJLZ-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Chlorthalidone related compound A USP reference standard for use in specified quality tests and assays. Also used to prepare chlorthalidone related compound A solution during the assay of chlorthalidone as described in United States Pharmacopeia.
Also, for use with USP monographs such as:
  • Chlorthalidone Tablets
  • Chlorthalidone

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2915BP0751A11040
description

United States Pharmacopeia (USP) Reference Standard

description

pharmaceutical secondary standard, certified reference material

description

British Pharmacopoeia (BP) Reference Standard

description

pharmaceutical analytical impurity (PAI)

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

BP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

pharmaceutical secondary standard, certified reference material

grade

pharmaceutical primary standard

grade

pharmaceutical analytical impurity (PAI)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical small molecule

format

neat

format

-

format

neat

format

neat

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

API family

chlorthalidone

API family

chlorthalidone

API family

chlorthalidone

API family

-


Still not finding the right product?

Explore all of our products under Chlorthalidone Related Compound A


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters