Skip to Content
Merck

1084292

USP

CD34+ Cell Enumeration System Suitability

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

form

lyophilized

manufacturer/tradename

USP

application(s)

USP Biologics
pharmaceutical

format

neat

storage temp.

−20°C

General description

The CD34+ Cell Enumeration System Suitability standard is designed for assessing the performance of analytical systems used in the enumeration of CD34+ hematopoietic stem and progenitor cells. This USP reference standard ensures accuracy and reproducibility in flow cytometry-based assays, which are crucial for evaluating stem cell doses in transplantation and cell therapy. It provides a consistent benchmark for laboratory systems, validating system suitability and operator performance in cell enumeration workflows.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

The USP CD34+ Cell Enumeration System Suitability Reference Standard comprises fixed and lyophilized human leukocytes, erythrocytes, and CD34+ cells derived from mobilized peripheral blood collected by apheresis of a G-CSF mobilized donor.

Application

CD34+ Cell Enumeration System Suitability reference standard intended for use in specified quality tests and assays.

Also, use to prepare system suitability standard solution during flow cytometric enumeration of cd34+ cells.

Additionally, used to assess the reagents and ensures correct gating during data acquisition and analysis, detection, and quantitation of CD34+ HSCs.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

This product is part of the USP Biologics program.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Ph
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia. The USP CD34+ Cell Enumeration System Suitability Reference Standard comprises fixed and lyophilized human leukocytes, erythrocytes, and CD34+ cells derived from mobilized peripheral blood collected by apheresis of a G-CSF mobilized donor.


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library