Skip to Content
Merck

Skip To

1064122

USP

N-Benzyl-N-methylethanolamine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-(N-Benzyl-N-methylamino)ethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
3 x 0.2 mL
Please contact Customer Service for Availability
CZK 30,900.00

About This Item

Linear Formula:
C6H5CH2N(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
165.23
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

CZK 30,900.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

nicardipine

manufacturer/tradename

USP

refractive index

n20/D 1.529 (lit.)

bp

95-105 °C/2 mmHg (lit.)

density

1.017 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CN(CCO)Cc1ccccc1

InChI

1S/C10H15NO/c1-11(7-8-12)9-10-5-3-2-4-6-10/h2-6,12H,7-9H2,1H3

InChI key

WOUANPHGFPAJCA-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

N-Benzyl-N-methylethanolamine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
36641210006991639003
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

USP

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

technical grade

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

neat

format

neat

API family

nicardipine

API family

-

API family

acebutolol

API family

sulfisoxazole acetyl

refractive index

n20/D 1.529 (lit.)

refractive index

n20/D 1.529 (lit.)

refractive index

-

refractive index

-


Still not finding the right product?

Explore all of our products under N-Benzyl-N-methylethanolamine


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters