Skip to Content
Merck

Skip To

1043149

USP

Ascorbyl Stearate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

L-Ascorbic acid 6-stearate, 6-O-Stearyl-L-ascorbic acid, L-Ascorbyl 6-octadecanoate, Ascorbyl stearate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
500 mg
Please contact Customer Service for Availability
5 160,00 CZK

About This Item

Empirical Formula (Hill Notation):
C24H42O7
CAS Number:
Molecular Weight:
442.59
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107

5 160,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

manufacturer/tradename

USP

application(s)

(Pharmaceutical small molecule)

format

neat

InChI

1S/C24H42O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(26)30-18-19(25)23-21(27)22(28)24(29)31-23/h19,23,25,28-29H,2-18H2,1H3

InChI key

NRHBEGWPKVFGRH-UHFFFAOYSA-N

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USP issued SDS can be found here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1043105BP4611086356
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

USP

application(s)

(Pharmaceutical small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

format

neat

format

neat

format

neat

format

neat

agency

USP

agency

-

agency

-

agency

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters