Skip to Content
Merck
All Photos(1)

Key Documents

TA9H97BAEC8A

Targetmol Chemicals Inc.

Xenopsin 2TFA(51827-01-1(free base)

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
1208.23

Iupac Name

((S)-5-oxopyrrolidine-2-carbonyl)glycyl-L-lysyl-L-arginyl-L-prolyl-L-tryptophyl-L-isoleucyl-L-leucine--2,2,2-trifluoroacetic acid (1/2)

Inchi Code

1S/C47H73N13O10.2C2HF3O2/c1-5-27(4)39(44(67)58-35(46(69)70)22-26(2)3)59-42(65)34(23-28-24-52-30-13-7-6-12-29(28)30)57-43(66)36-16-11-21-60(36)45(68)33(15-10-20-51-47(49)50)56-41(64)31(14-8-9-19-48)55-38(62)25-53-40(63)32-17-18-37(61)54-32;2*3-2(4,5)1(6)7/h6-7,12-13,24,26-27,31-36,39,52H,5,8-11,14-23,25,48H2,1-4H3,(H,53,63)(H,54,61)(H,55,62)(H,56,64)(H,57,66)(H,58,67)(H,59,65)(H,69,70)(H4,49,50,51);2*(H,6,7)/t27-,31-,32-,33-,34-,35-,36-,39-;;/m0../s1

InChI key

UCLPMNPCTKAOLM-XHOQWZSJSA-N

storage temp.

-80/-20

Purity

100%

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

Product Link

https://www.targetmol.com/compound/Xenopsin 2TFA(51827-01-1(free base)

MSDS

https://www.targetmol.com/compound/Xenopsin 2TFA(51827-01-1(free base)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library