Skip to Content
Merck
All Photos(1)

Key Documents

Y0002194

Pentobarbital sodium

CRS, European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pentobarbital sodium salt, 5-Ethyl-5-(1-methylbutyl)-2,4,6-trioxohexahydropyrimidine, Nembutal

Sign Into View Organizational & Contract Pricing

Select a Size

40 MG
CZK 2,770.00

CZK 2,770.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
40 MG
CZK 2,770.00

About This Item

Empirical Formula (Hill Notation):
C11H17N2NaO3
CAS Number:
Molecular Weight:
248.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 2,770.00


Please contact Customer Service for Availability

Request a Bulk Order

API family

pentobarbital

manufacturer/tradename

EDQM

storage temp.

2-8°C

SMILES string

[Na].CCCC(C)C1(CC)C(=O)NC(=O)NC1=O

InChI

1S/C11H18N2O3.Na.H/c1-4-6-7(3)11(5-2)8(14)12-10(16)13-9(11)15;;/h7H,4-6H2,1-3H3,(H2,12,13,14,15,16);;

InChI key

PGOYUXGHADTNIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pentobarbital sodium EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. sous quelle forme est ce produit ?

    1 answer
    1. Product number Y0002194 (Pentobarbital sodium) is a powder.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service