SU860155
Short thread screw cap, 9 mm thread, with liner
UltraBond, blue polypropylene screw cap (open-top)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121806
NACRES:
NB.11
material
beige silicon liner
blue polypropylene screw cap (open-top)
feature
UltraBond
hardness
45 (Durometer: shore A)
packaging
pkg of 1 × 1000 ea (SU860155)
availability
not available in USA
thickness
1.3 mm
fitting
for 9 mm (short thread)
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service