Skip to Content
Merck
All Photos(1)

Key Documents

SPRF7

Smoke Point Reference Fuel Blend

Fuel Blend 7 for 42.8 mm (0/100%v/v), certified reference material

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CZK 3,650.00

CZK 3,650.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
CZK 3,650.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

CZK 3,650.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service