Skip to Content
Merck
All Photos(1)

Key Documents

SAAR6018250

Thymol

uniLAB

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907135
MDL number:
UNSPSC Code:
41116100

vapor pressure

1 mmHg ( 64 °C)

product line

uniLAB

packaging

pkg of 100 g (SAAR6018250DC)

bp

232 °C (lit.)

mp

48-51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

InChI key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service