Skip to Content
Merck
All Photos(1)

Key Documents

PS672

Prophos

analytical standard

Synonym(s):

Ethoprophos, O-Ethyl S,S-dipropyl phosphorodithioate, Ethoprofos, Ethoprop

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19O2PS2
CAS Number:
Molecular Weight:
242.34
Beilstein:
8139788
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-672

application(s)

agriculture
environmental

SMILES string

CCCSP(=O)(OCC)SCCC

InChI

1S/C8H19O2PS2/c1-4-7-12-11(9,10-6-3)13-8-5-2/h4-8H2,1-3H3

InChI key

VJYFKVYYMZPMAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prophos is an insecticide and nematicide, which is incorporated into the soil in granular formulations. It is used in controlling parasitic nematodes in plants and soil insects. Its mode of action basically involves the inhibition of acetylcholinesterase.

Application

Prophos may be used as a reference standard in the determination of prophos in samples of agrochemicals using direct analysis in real time ionization coupled to a single-quadrupole mass spectrometer (DART-MS).

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct analysis in real time mass spectrometry for the rapid identification of four highly hazardous pesticides in agrochemicals.
Wang L, et al.
Rapid Communications in Mass Spectrometry, 26(16), 1859-1867 (2012)
Metabolic Pathways of Agrochemicals: Part 2: Insecticides and Fungicides (2007)
S D Ray et al.
Research communications in molecular pathology and pharmacology, 107(1-2), 105-128 (2001-05-04)
Several observations, both in humans and laboratory animals, have suggested that proanthocyanidins exhibit a broad spectrum of pharmacological, therapeutic and chemoprotective properties. Specifically, some of our earlier studies have shown that IH636 grape seed proanthocyanidin extract (GSPE, commercially known as
Safety related to exposure: dermal dose-red cell cholinesterase response relationships for ethoprop and Mocap 6EC.
J B Knaak et al.
Bulletin of environmental contamination and toxicology, 38(5), 834-839 (1987-05-01)
D E Robinson et al.
The Science of the total environment, 237-238, 373-378 (1999-11-24)
The hydrolytic half lives of ethoprophos in distilled, river, brackish and open sea water were 25, 133, 65 and 81 days, respectively. Under laboratory conditions, volatilisation of the residues after 12 h was 1.4-3.6, 2.3-4.5 and 6.5-20.2% from a sandy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service