Skip to Content
Merck
All Photos(1)

Documents

PS605

Dibrom®

analytical standard

Synonym(s):

(1,2-Dibromo-2,2-dichloroethyl) dimethyl phosphate, Bromchlophos, Naled

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7Br2Cl2O4P
CAS Number:
Molecular Weight:
380.78
Beilstein:
2049930
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 250 mg

manufacturer/tradename

Chem Service, Inc. PS-605

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COP(=O)(OC)OC(Br)C(Cl)(Cl)Br

InChI

1S/C4H7Br2Cl2O4P/c1-10-13(9,11-2)12-3(5)4(6,7)8/h3H,1-2H3

InChI key

BUYMVQAILCEWRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Dibrom is a registered trademark of AMVAC Chemical Corporation

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiu Gan et al.
Environmental science & technology, 40(18), 5717-5723 (2006-09-30)
Reactions of dichlorvos with five reduced sulfur species (hydrogen sulfide, bisulfide, thiosulfate, thiophenol, and thiophenolate) were examined in well-defined anoxic aqueous solutions to investigate their role in its degradation. Reactions were monitored at varying concentrations of reduced sulfur species over
Timothy A Bargar
Environmental toxicology and chemistry, 31(4), 885-891 (2012-01-27)
A prospective risk assessment was conducted for adult butterflies potentially exposed to the mosquito control insecticide naled. Published acute mortality data, exposure data collected during field studies, and morphometric data (total surface area and fresh body weight) for adult butterflies
Chi-Chun Fang et al.
Journal of economic entomology, 104(6), 2039-2045 (2012-02-04)
Naled is a commonly used insecticide for controlling populations of the oriental fruit fly, Bactrocera dorsalis (Hendel), in Taiwan and other countries. B. dorsalis has developed resistance to the insecticide, and the resistance management is an important issue. Ecological effects
Mark S Breidenbaugh et al.
Journal of the American Mosquito Control Association, 24(2), 327-330 (2008-08-01)
Post-Hurricane Rita mosquito surveillance was carried out in 4 east Texas counties to determine mosquito abundance, species composition, and need for mosquito control. Subsequently, aerial applications of naled (Dibrom) for mosquito control were made by the Air Force Aerial Spray
Ryan S Davis et al.
Integrated environmental assessment and management, 3(3), 373-382 (2007-08-19)
West Nile virus (WNV) has been a concern for people across the United States since the disease was initially observed in the summer of 1999. Since 1999, WNV has caused the largest arboviral encephalitis epidemic in US history. Vector control

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service