Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C13H14F3N3O4
CAS Number:
Molecular Weight:
333.26
EC Number:
259-564-3
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
2907084
MDL number:
grade
analytical standard
packaging
ampule of 1000 mg
manufacturer/tradename
Chem Service, Inc. PS-408
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
agriculture
environmental
format
neat
SMILES string
CCN(CC(C)=C)c1c(cc(cc1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O
InChI
1S/C13H14F3N3O4/c1-4-17(7-8(2)3)12-10(18(20)21)5-9(13(14,15)16)6-11(12)19(22)23/h5-6H,2,4,7H2,1,3H3
InChI key
PTFJIKYUEPWBMS-UHFFFAOYSA-N
General description
Ethalfluralin belongs to a class of methyl dinitroaniline herbicide which is mainly effective against broad-leaved weeds and annual grasses.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ethalfluralin may be used as a reference standard for the determination of ethalfluralin in water sample by high-performance liquid chromatography post solid-phase extraction.
Still not finding the right product?
Explore all of our products under Ethalfluralin
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


