Skip to Content
Merck

PS1031

Fentin hydroxide

analytical standard

Synonym(s):

Triphenyltin hydroxide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C18H16OSn
CAS Number:
Molecular Weight:
367.03
EC Number:
200-990-6
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
4139186
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

analytical standard

packaging

ampule of 500 mg

manufacturer/tradename

Chem Service, Inc. PS-1031

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

SMILES string

O[Sn](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/3C6H5.H2O.Sn/c3*1-2-4-6-5-3-1;;/h3*1-5H;1H2;/q;;;;+1/p-1

InChI key

BFWMWWXRWVJXSE-UHFFFAOYSA-M

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Species differences in the sensitivity to the diabetogenic action of triphenyltin hydroxide].
H Matsui et al.
Sangyo igaku. Japanese journal of industrial health, 25(3), 190-191 (1983-05-01)
Marcia Sarpa et al.
Regulatory toxicology and pharmacology : RTP, 49(1), 43-52 (2007-07-10)
Triphenyltin-hydroxide (TPTH) is used as agricultural fungicide in Brazil and elsewhere. This study was undertaken to evaluate the developmental toxicity of TPTH in mice. Swiss Webster mice were treated by gavage with TPTH (0, 3.75, 7.5, 15 and 30 mg/kg
David I Shapiro-Ilan et al.
Environmental entomology, 40(1), 59-65 (2011-12-21)
Hypocreales fungi such as Beauveria bassiana (Balsamo) Vuillemin and Metarhizium brunneum Petch can be negatively affected by fungicides thereby reducing their biocontrol potential. In a previous study, we demonstrated enhanced fungicide resistance in B. bassiana through artificial selection. However, it
S Tanimoto et al.
Molecular reproduction and development, 39(4), 409-414 (1994-12-01)
Flux of K+ and changes in intracellular Ca2+ in the sperm of salmonid fishes were measured with spectrophotometry, ion electrode, microscopic fluorometry, and radioisotope accumulation. Release of K+ occurred at the initiation of sperm motility which is induced by decrease
Eduardo C Oliveira-Filho et al.
Chemosphere, 81(2), 218-227 (2010-07-03)
Plant molluscicides have been regarded as possible alternatives to the costly and environmentally hazardous molluscicides currently available. This study was undertaken to compare the developmental toxicity of a plant molluscicide (Euphorbia milii latex, LAT) with that of three synthetic molluscicidal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service