K08670686
Kromasil® C4 HPLC Column
10 μm particle size, pore size 100 Å, L × I.D. 150 mm × 4.6 mm
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Agency
suitable for USP L26
product line
Kromasil®
manufacturer/tradename
Kromasil® KR100-10-C4-4.6X150
availability
available only in USA, Canada and Puerto Rico
available only in USA, Canada and Puerto Rico
technique(s)
HPLC: suitable
L × I.D.
150 mm × 4.6 mm
matrix active group
C4 (butyl) phase
particle size
10 μm
pore size
100 Å
application(s)
food and beverages
separation technique
reversed phase
Looking for similar products? Visit Product Comparison Guide
Legal Information
Kromasil is a registered trademark of EKA Chemicals AB
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service