G005951
HPLC Analysis of Fusion Protein on Zenix®-C SEC-300 vs. Zenix® SEC-300
application for HPLC
About This Item
Recommended Products
technique(s)
HPLC: suitable
test parameters
column: Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777033)
column: Zenix-C SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777092)
mobile phase: 150 mM sodium phosphate buffer, pH 7.0 + 200 mM NaCl
flow rate: 0.5 mL/min
column temp.: 25 °C
detector: UV, 214 nm
injection: 10 μL
sample: 1 mg/mL fusion protein, MW 170 kD, pl 6.8-7.0
suitability
application for HPLC
application(s)
life science and biopharma
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service