Skip to Content
Merck
All Photos(1)

Key Documents

G005375

HPLC Analysis of 25-Dihydroxyvitamin D2, 25-Hydroxyvitamin D3 and 3-epi-25-Hydroxyvitamin D3 on Ascentis® Express F5, 20 °C

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Ascentis Express F5, 10 cm x 2.1 mm I.D., 2.7 μm particles (53569-U)
mobile phase: [A] 7 mM ammonium formate; [B] methanol (25:75)
flow rate: 0.4 mL/min
pressure: 5221 psi (245 bar)
column temp.: 20 °C
detector: (UV, 265 nm, ESI(+), m/z 100 - 1000)
injection: 1 μL
sample: 300 mg/L in 25:75, water:methanol

suitability

application for HPLC

application(s)

clinical
vitamins, nutraceuticals, and natural products

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    25-Hydroxyvitamin D2 solution, 50 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
    25-Hydroxyvitamin D3 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
    3-epi-25-Hydroxyvitamin D3, 98% (CP)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service