G004683
LC/MS Analysis of Ancymidol Enantiomers on Astec® CYCLOBOND I 2000 HP-RSP
application for HPLC
About This Item
technique(s)
HPLC: suitable
test parameters
column: CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 μm particles (24024AST)
mobile phase: [A] 5 mM ammonium acetate, pH 6.0; [B] acetonitrile; (80:20, A:B)
flow rate: 1 mL/min
column temp.: 35 °C
detector: MS (m/z = 257)
injection: 5 μL
sample: 1 mg/mL in methanol
suitability
application for HPLC
application(s)
agriculture
Legal Information
Analyte
analytical column
standard
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service