G003908
GC Analysis of Fuel Oil #6 on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
suitable for GC
About This Item
technique(s)
gas chromatography (GC): suitable
test parameters
column: Equity-1, 15 m x 0.10 mm I.D., 0.10 μm (28039-U)
oven: 80 °C, 50 °C/min to 325 °C
inj. temp.: 250 °C
detector: FID, 350 °C
carrier gas: hydrogen, 45 cm/sec constant
injection: 0.3 μL, 100:1 split, 0.1 min pre-injection dwell time
liner: 2 mm I.D., straight
sample: No.6 Fuel Oil standard, 20 mg/mL in hexane:chloroform (47536-U)
application(s)
petroleum
Legal Information
analytical column
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service