G000595
HPLC Analysis of Tricyclic Antidepressants in Serum on Discovery® C18 after SPE using Discovery® DSC-18
application for SPE, application for HPLC
About This Item
technique(s)
HPLC: suitable
solid phase extraction (SPE): suitable
test parameters
sample preparation: SPE (Solid Phase Extraction)
SPE tube/cartridge: Discovery DSC-18, 100 mg/1 mL (52602-U)
column: Discovery C18, 15 cm × 4.6 mm, 5 μm preceded by a 2 cm C18 guard column and 0.5 μm frit filter (504955)
mobile phase: [A] acetonitrile: [B] methanol: [C] 25 mM potassium phosphate, monobasic (pH 7 with triethlylamine) (45:25:30, A:B:C)
flow rate: 1 mL/min
column temp.: ambient
detector: UV, 254 nm
injection: 50 μL, diluted porcine serum extract
suitability
application for HPLC
application for SPE
application(s)
clinical
Legal Information
Analyte
analytical column
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service