Skip to Content
Merck
All Photos(1)

Key Documents

ES5364

GreenSep Pyridyl Amide SFC 5 Micron HPLC Column

5 μm particle size, L × I.D. 25 cm × 30 mm

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 210,800.00

CZK 210,800.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
CZK 210,800.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

CZK 210,800.00


Please contact Customer Service for Availability

material

stainless steel column

Quality Level

product line

GreenSep

manufacturer/tradename

ES Industries 15N291-GS-PYA

packaging

1 ea of

technique(s)

HPLC: suitable

L × I.D.

25 cm × 30 mm

matrix

fully porous particle

matrix active group

pyridyl amide phase

particle size

5 μm

pore size

120 Å pore size

operating pH

2-8

separation technique

supercritical fluid (SFC)

Looking for similar products? Visit Product Comparison Guide

General description

A metal free, ultra high purity chromatographic media that is pressure stable and specifically engineered for high performance SFC separations. The surface is treated to produce optimum SFC separation interactions and loading capacity while maintaining superior peak shape perfor- mance for many chemicals. GreenSep Silica is available for analytical and preparative column formats in particle sizes from 1.8 μm to 20 μm.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

GreenSep is a trademark of ES Industries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service