Skip to Content
Merck
All Photos(1)

Key Documents

CRM34311

Supelco

Acifluorfen

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

5-(2-Chloro-α,α,α-trifluoro-p-tolyloxy)-2-nitrobenzoic acid, 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H7ClF3NO5
CAS Number:
Molecular Weight:
361.66
Beilstein:
2953865
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

−20°C

SMILES string

OC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+]([O-])=O

InChI

1S/C14H7ClF3NO5/c15-10-5-7(14(16,17)18)1-4-12(10)24-8-2-3-11(19(22)23)9(6-8)13(20)21/h1-6H,(H,20,21)

InChI key

NUFNQYOELLVIPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

Acifluorfen is a post-emergence herbicide and particularly effective against broad-leaved weeds and grasses. It is also a pesticide transformation product. As per the regulation (EC) No 1107/2009 it is not approved for use as plant protection product in the European Union. A default maximum residuel level (MRL) of 0.01 mg/kg has been set for acifluorfen according to Art 18(1)(b)Reg. (EU) No 2022/1346 for various products of plant and animal origin from 0.05 - 15 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Environmental

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service