Skip to Content
Merck
All Photos(1)

Key Documents

95659

Supelco

1,7-Octadiene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH(CH2)4CH=CH2
CAS Number:
Molecular Weight:
110.20
Beilstein:
605288
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.21
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

technique(s)

HPLC: suitable

refractive index

n20/D 1.422 (lit.)

bp

114-121 °C (lit.)

density

0.746 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-30°C

SMILES string

C=CCCCCC=C

InChI

1S/C8H14/c1-3-5-7-8-6-4-2/h3-4H,1-2,5-8H2

InChI key

XWJBRBSPAODJER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service