Skip to Content
Merck
All Photos(1)

Key Documents

842176

TSKgel® Super-Octyl (2.3 µm) HPLC Columns

L x I.D. 1 cm × 2 mm, C8 (octyl) phase,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® Super-Octyl Guard Cartridge, phase C8 (octyl), L × I.D. 1 cm × 2 mm, 2.3 μm particle size

material

stainless steel

Agency

suitable for USP L7

product line

TSKgel®

manufacturer/tradename

Tosoh 42176

extent of labeling

5% Carbon, multilayer, endcapped

parameter

no limitation salt concentration
0-100% organic solvent
10-50 °C temperature

technique(s)

HPLC: suitable

L × I.D.

1 cm × 2 mm

surface area

100 m2/g

matrix

spherical silica

matrix active group

C8 (octyl) phase,

particle size

2.3 μm

operating pH

2.0-7.5

separation technique

reversed phase

General description

TSKgel® Guard cartridges require a holder, which is sold separately. For 2 mm cartridges, the holder is Part No. 819308. For the 3.2 mm cartridges used to protect 4.6 mm columns, the holder is Part No. 819018.
_x000D_
TSKgel® Super-Octyl Guard Cartridge, 3pk, for 2mmID TSKgel Super-Octyl columns.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service