Skip to Content
Merck
All Photos(1)

Key Documents

842102

TOYOPEARL® AF-Red-650ML Bulk Media

phase Procion Red HE-3B, 65 μm particle size, bottle of 1 L

Synonym(s):

TOYOPEARL® AF-Red-650M Bulk Media

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
CZK 148,800.00

CZK 148,800.00


Please contact Customer Service for Availability


Select a Size

Change View
1 L
CZK 148,800.00

About This Item

UNSPSC Code:
41106500

CZK 148,800.00


Please contact Customer Service for Availability

description

Adsorption capacity (3.5 +/- 1.0 mg/mL (human serum albumin))
Phase filter (Red)

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 42102

extent of labeling

7 μmol/mL ligand density

parameter

≤3 bar pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer particle platform

matrix active group

Procion Red HE-3B phase

particle size

65 μm

pore size

1000 Å

pH

4-9

capacity

3.5 g/L±1.0 adsorption capacity (human serum albumin)

separation technique

affinity

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl AF-Red-650 resin is functionalized with Procion Red HE-3B (also known as Reactive Red 120). It is used for nucleotide-dependent enzymes, lipoproteins, plasminogen, peptides, hormones, and cytotoxins

Physical form

72% (v/v) slurry in 20% (v/v) ethanol

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service