Skip to Content
Merck
All Photos(1)

Key Documents

821966

TSKgel® CM-STAT (7 µm) HPLC Columns

L x I.D. 10 cm × 4.6 mm, carboxymethyl phase

Synonym(s):

TSKgel® CM-STAT Columns

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 41,400.00

CZK 41,400.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
CZK 41,400.00

About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

CZK 41,400.00


Please contact Customer Service for Availability

Product Name

TSKgel® CM-STAT HPLC Column, phase carboxymethyl, L × I.D. 10 cm × 4.6 mm, 7 μm particle size

material

stainless steel hardware

product line

TSKgel®

feature

counter ion Na+

manufacturer/tradename

Tosoh CM-STAT, 21966

parameter

no limitation salt concentration
≤50% organic solvent
1.0-2.0 mL/min standard flow rate
10-60 °C temperature
100 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

10 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

carboxymethyl phase

particle size

7 μm

operating pH

3-10

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Bulk packing to top off and/or repack TSKgel columns and guard columns.
TSKgel CM-STAT columns are packed with 7 and 10μm hydrophilic non-porous resin particles of which the surface consists of an open access network of multi-layered weak cation exchange groups. The innovative bonding chemistry, combined with a relatively large particle size, results in a respectable loading capacity, low operating pressure and rapid analysis. Applications for the TSKgel CM-STAT columns include the separation of proteins, protein aggregates, charge isomers of monoclonal antibodies, PEGylated proteins, and peptide digests.

Physical form

Shipped in DI water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service