Skip to Content
Merck
All Photos(1)

Key Documents

819808

TOYOPEARL® HW-40F Bulk Media

phase hydroxylated methacrylate, bottle of 150 mL, 45 μm particle size

Synonym(s):

TOYOPEARL® HW-40 Size Exclusion Media

Sign Into View Organizational & Contract Pricing

Select a Size

150 ML
CZK 7,540.00

CZK 7,540.00


Please contact Customer Service for Availability


Select a Size

Change View
150 ML
CZK 7,540.00

About This Item

UNSPSC Code:
41106500

CZK 7,540.00


Please contact Customer Service for Availability

product line

TOYOPEARL®

form

slurry

packaging

bottle of 150 mL

manufacturer/tradename

Tosoh 19808

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylate phase

particle size

45 μm

operating pH

2-13

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl HW-40 resin is a hydroxylated methacrylic polymer for size exclusion chromatography. It is typically used for high throughput desalting and buffer exchange. It also can remove surfactants such as Triton X100 from protein solutions. It is often used for smaller biomolecules with exclusion limits up to 10,000 Da.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service