Skip to Content
Merck
All Photos(1)

Documents

808800

TSKgel® G3000SW (10 µm) HPLC Columns

L x I.D. 30 cm × 8 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

product name

TSKgel® G3000SW HPLC Column, phase diol, L × I.D. 30 cm × 8 mm, 10 μm particle size, glass

material

glass

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 08800

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
0.8 mL/min flow rate
10-30 °C temperature
20 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 8 mm

matrix

spherical silica

matrix active group

diol phase

particle size

10 μm

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service