Skip to Content
Merck

Skip To

808043

TSKgel® Phenyl-5PW RP (10 µm) HPLC Columns

L x I.D. 7.5 cm × 4.6 mm, phenyl phase

Synonym(s):

TSKgel® Reversed Phase HPLC Column (Polymer-based)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
30 100,00 CZK

About This Item

UNSPSC Code:
41115700
NACRES:
NB.21
L × i.d.:
7.5 cm × 4.6 mm
Particle size:
10 μm
Matrix active group:
phenyl phase
Pore size:
1000 Å pore diameter
Matrix:
polymer particle platform

30 100,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

TSKgel® Phenyl-5PW RP HPLC Column, phase phenyl, L × I.D. 7.5 cm × 4.6 mm, 10 μm particle size

material

stainless steel

product line

TSKgel®

mol wt

1000 - 1,000,000

manufacturer/tradename

Tosoh Phenyl-5PW, 08043

extent of labeling

1 meq/mL ligand density (estimate)

parameter

no limitation salt concentration, 0-100% organic solvent, 1.2 mL/min flow rate, 30 bar max. pressure, 4-60 °C temperature

technique(s)

HPLC: suitable

L × I.D.

7.5 cm × 4.6 mm

matrix

polymer particle platform

matrix active group

phenyl phase

particle size

10 μm

pore size

1000 Å pore diameter

operating pH

2-12

separation technique

reversed phase

General description

Polymer-based particles for reversed-phase (RP) HPLC generally have two benefits over silica-based particles: They are more tolerant of high and low pH operating conditions and they do not have the silanol interactions that cause peak tailing of some basic analytes. Also, some silicas have an intrinsic metal content that may cause chelation problems with sensitive biomolecules, leading to poor recovery. Polymeric TSKgel columns come in C18 and phenyl functional groups and a range of particle porosities. Fast kinetics and quantitative protein recovery at submicrogram loading are characteristic of nonporous resin-based TSKgel Octadecyl-NPR columns.
  • Octadecyl-2PW particles have 125Å pores
  • Octadecyl-4PW particles have 500Å pores
  • Phenyl-5PW RP particles have 1000Å pores
  • Octadecyl-NPR particles are nonporous
TSKgel Phenyl-5PW RP columns are filled with 10 micron porous polymethacrylate particles, the surface of which is covered with a high density of phenyl groups that causes proteins to be retained in a low salt buffer mobile phase and requiring a solvent gradient for their elution, as is the protocol in reversed phase chromatography. In contrast, TSKgel Phenyl-5PW has such a low density of phenyl groups on the surface that proteins can only be retained at high salt concentration and eluted by a decreasing salt gradient, which, of course, is the protocol used in hydrophobic interaction chromatography. TSKgel Phenyl-5PW RP columns can withstand rigorous cleaning with either acid or base. The 1000 A pore size accommodates globular protein samples up to 1,000,000 Da.

Physical form

Shipped in 30% methanol - 70% water.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
814007818756817500
description

phase phenyl, L × I.D. 7.5 cm × 4.6 mm, 10 μm particle size

description

phase phenyl, octadecyl, L × I.D. 7.5 cm × 8 mm, 10 μm particle size

description

phase phenyl, L × I.D. 7.5 cm × 2 mm, 10 μm particle size

description

phase C18 (octadecyl), L × I.D. 15 cm × 4.6 mm, 5 μm particle size

separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

separation technique

reversed phase

matrix active group

phenyl phase

matrix active group

phenyl phase, octadecyl

matrix active group

phenyl phase

matrix active group

C18 (octadecyl) phase

L × I.D.

7.5 cm × 4.6 mm

L × I.D.

7.5 cm × 8 mm

L × I.D.

7.5 cm × 2 mm

L × I.D.

15 cm × 4.6 mm

particle size

10 μm

particle size

10 μm

particle size

10 μm

particle size

5 μm

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable

product line

TSKgel®

product line

TSKgel®

product line

TSKgel®

product line

TSKgel®


Still not finding the right product?

Explore all of our products under TSKgel® Phenyl-5PW RP HPLC Column




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters