Skip to Content
Merck
All Photos(1)

Key Documents

80445

Supelco

Kynurenic acid-4a,5,6,7,8,8a-13C6

analytical standard

Synonym(s):

4-Hydroxy-2-quinoline-4a,5,6,7,8,8a-13C6-carboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

80445-10MG
CZK 7,510.00

CZK 7,510.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
80445-10MG
CZK 7,510.00

About This Item

Empirical Formula (Hill Notation):
13C6C4H7NO3
Molecular Weight:
195.12
UNSPSC Code:
41116107
NACRES:
NA.21

CZK 7,510.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service