Skip to Content
Merck
All Photos(2)

Key Documents

74477

Supelco

o-Phenylenediamine

analytical standard

Synonym(s):

1,2-Diaminobenzene, 1,2-Phenylenediamine, OPD

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
CZK 1,570.00

CZK 1,570.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
CZK 1,570.00

About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
Beilstein:
606074
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

CZK 1,570.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

3.7 (vs air)

vapor pressure

0.01 mmHg ( 25 °C)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

256-258 °C

format

neat

storage temp.

2-30°C

SMILES string

Nc1ccccc1N

InChI

1S/C6H8N2/c7-5-3-1-2-4-6(5)8/h1-4H,7-8H2

InChI key

GEYOCULIXLDCMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

276.8 °F - closed cup

Flash Point(C)

136 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service