Skip to Content
Merck
All Photos(1)

Key Documents

69563

Supelco

N-Acetylglycine

analytical standard

Synonym(s):

Acetamidoacetic acid, Aceturic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH2CO2H
CAS Number:
Molecular Weight:
117.10
Beilstein:
774114
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Assay

≥95.0% (HPLC)
98.0-102.0% (T)

form

powder

shelf life

limited shelf life, expiry date on the label

mp

207-209 °C (lit.)

application(s)

clinical testing

storage temp.

2-8°C

SMILES string

CC(=O)NCC(O)=O

InChI

1S/C4H7NO3/c1-3(6)5-2-4(7)8/h2H2,1H3,(H,5,6)(H,7,8)

InChI key

OKJIRPAQVSHGFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N-Acetylglycine, belongs to the class of α-amino acids, which reportedly create stable salts when combined with organic bases.

Application

N-Acetylglycine may be used as an internal standard for the determination of N-acetylneuraminic acid and N- glycolylneuraminic acid in glycoproteins by high- performance liquid chromatography (LC) with UV detection.
It may also be used as an analytical standard:
  • For studying the nontargeted modification-specific metabolomics based on liquid chromatography with high-resolution mass spectrometry (LC-HRMS).
  • Determination of the analyte in tomato and potato samples using ion-pair liquid chromatography coupled to mass spectrometry (IP-LC-MS).

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nontargeted modification-specific metabolomics study based on liquid chromatography?high-resolution mass spectrometry
Dai W, et al.
Analytical Chemistry, 86(18), 9146-9153 (2014)
Determination of N-acetylneuraminic acid and N-glycolylneuraminic acid in glycoproteins by high-performance liquid chromatography without derivatization
Ogawa KH, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 612(1), 145-149 (1993)
Academic Press Dictionary of Science and Technology, 76 (1992)
Calorimetric determination of the heats of formation of competitive ternary mixed-ligand complex compounds. copper (II)-N-acetylglycine-?-amino ac
Thermochimica Acta, 123, 201-212 (1988)
Comprehensive Analysis of Umami Compounds by Ion?Pair Liquid Chromatography Coupled to Mass Spectrometry
Coulier L, et al.
Journal of Food Science, 76(7), C1081-C1087 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service