Skip to Content
Merck
All Photos(1)

Key Documents

68429

Supelco

N-Methyl-N-nitroso-2-phenylethylamine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

N-Methyl-N-nitrosobenzeneethanamine, N-Methyl-N-nitrosophenethylamine, N-Nitroso-N-methyl-2-phenylethylamine, Nitroso-N-methyl-N-(2-phenyl)ethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
CZK 2,690.00

CZK 2,690.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
CZK 2,690.00

About This Item

Empirical Formula (Hill Notation):
C9H12N2O
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.21

CZK 2,690.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

storage temp.

−20°C

SMILES string

N(N=O)(CCc1ccccc1)C

InChI key

AVESHLVLHJKUKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service