Skip to Content
Merck
All Photos(1)

Documents

58217

Supelco

Column Blank Kit for HPLC

I.D. × O.D. × L 10 mm × 1/2 in. × 25 cm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

I.D. × O.D. × L

10 mm × 1/2 in. × 25 cm

Looking for similar products? Visit Product Comparison Guide

General description

Each column blank kit includes a precision-bore polished 316 stainless steel column blank, two zero dead volume external reducing unions (internal unions with 10 mm I.D. blanks) with nuts and ferrules for 1/16 in. O.D. tubing, and two
2 μm frits.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service