Skip to Content
Merck
All Photos(1)

Key Documents

57545

Low Volume Static Mixer (50-250 μL)

50 μL cartridge, stainless steel

Synonym(s):

50UL STATIC MIXER CARTRIDGE

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 5,150.00

CZK 5,150.00

List PriceCZK 10,300.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
CZK 5,150.00

About This Item

UNSPSC Code:
41115854

CZK 5,150.00

List PriceCZK 10,300.00Save 50%

Please contact Customer Service for Availability

Request a Bulk Order

material

stainless steel

compatibility

for use with ASI low volume mixer housing

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service