Skip to Content
Merck

Skip To

57461

Rheodyne® Model 3725i Preparative Sample Injector

PEEK

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
51 000,00 CZK

About This Item

UNSPSC Code:
41115854
NACRES:
NB.21

51 000,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

PEEK

feature

manual, number of ports 6, positions 2

packaging

pkg of 1 ea

General description

Model 3725i sample injectors for 1-10 cm I.D. HPLC columns combine the ease of use and versatility of Rheodyne′s analytical-scale injectors with the ability to handle large samples and high flow rates. Rugged and easily maintained, in a choice of stainless steel or PEEK flowpaths. Includes 10 mL sample loop, 1/8 in. fittings for all ports, 5cm large-bore syringe needle (16 gauge/0.65 in. OD) for rapid injection of large samples, needle port cleaner, two vent tubes, wrenches, mounting screws, and instructions.

Other Notes

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
588275762157630
description

PEEK

description

-

description

Rheodyne 7725i

description

volume 1 mL

material

PEEK

material

-

material

-

material

stainless steel sample loop and fittings

packaging

pkg of 1 ea

packaging

-

packaging

-

packaging

-

feature

manual, positions 2, number of ports 6

feature

positions 2, number of ports 6

feature

positions 2, manual, number of ports 6

feature

-


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters